Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-N-methyl-D-phenylalanine, 98%

Chemical Identifiers

Chemical Name or Material N-Fmoc-N-methyl-D-phenylalanine
CAS 138775-05-0
Formula Weight 401.45
MDL Number MFCD00153392
Beilstein 8441627
Synonym Fmoc-N-Me-D-Phe-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6305503

 
alfa aesar™
H6305503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6305506

 
alfa aesar™
H6305506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63055MD

 
alfa aesar™
H63055MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 401.45
Chemical Name or Material N-Fmoc-N-methyl-D-phenylalanine
Physical Form Solid
Assay 98%
MDL Number MFCD00153392
CAS 138775-05-0
Beilstein 8441627
Synonym Fmoc-N-Me-D-Phe-OH
Quantity 1g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C