missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-N-methyl-D-leucine, 97%

Manufacturer:  Alfa Aesar™ H6389514

 View more versions of this product

Catalog No. AAH6389514



Description

Description

Applications
Tripeptides were prepared by means of coupling of N-Fmoc-N-methyl-D-leucine.

Solubility
Soluble in clear solution (0.3 gram in 2 ml DMF).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Specifications

Specifications

N-Fmoc-N-methyl-D-leucine
C22H25NO4
7350417
BUJQSIPFDWLNDC-HXUWFJFHSA-N
(2R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-4-methylpentanoic acid
7019520
Powder
White
103478-63-3
MFCD00235877
fmoc-n-me-d-leu-oh, fmoc-n-methyl-d-leucine, fmoc-d-n-me-leu-oh, d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2r-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-4-methyl-pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-4-methylpentanoic acid, fmoc-d-meleu-oh, ambotzfaa1517, pubchem18971, h-n-me-d-ala.hcl
CC(C)CC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
367.445
367.44
97%
25g

Chemical Identifiers

103478-63-3
367.445
BUJQSIPFDWLNDC-HXUWFJFHSA-N
7019520
CC(C)CC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C22H25NO4
MFCD00235877
fmoc-n-me-d-leu-oh, fmoc-n-methyl-d-leucine, fmoc-d-n-me-leu-oh, d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2r-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino-4-methyl-pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino-4-methylpentanoic acid, fmoc-d-meleu-oh, ambotzfaa1517, pubchem18971, h-n-me-d-ala.hcl
(2R)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]-4-methylpentanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents