missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-α-FMOC-N-epsilon-BOC-L-lysine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC265220250

 View more versions of this product

Catalog No. AC265220250



Description

Description

Specifications

Specifications

N-α-FMOC-N-ε-BOC-L-lysine
Glass bottle
− 12.00
129.0°C to 135.0°C
71989-26-9
100.0
C26H32N2O6
a-boc-l-lysine, a-fmoc-n, fmoc-lys boc, fmoc-lys boc-oh, n, n-alpha-fmoc-n-epsilon-tert-boc-l-lysine, n-alpha-fmoc-nepsilon-boc-l-lysine, n-fmoc-n'-boc-l-lysine, na-fmoc-ne-boc-l-lysine, nepsilon-boc-nalpha-fmoc-l-lysine, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid
CC(C)(C)OC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
468.54
468.54
98%
Authentic
−12° (24°C c=2,DMF)
White
25g
97.5
97.5% min. (HPLC)
FMOC-NHCH(COOH)(CH2)4NHC(O)OC(CH3)3
UMRUUWFGLGNQLI-QFIPXVFZSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
2724628
Powder

Chemical Identifiers

71989-26-9
468.54
a-boc-l-lysine, a-fmoc-n, fmoc-lys boc, fmoc-lys boc-oh, n, n-alpha-fmoc-n-epsilon-tert-boc-l-lysine, n-alpha-fmoc-nepsilon-boc-l-lysine, n-fmoc-n'-boc-l-lysine, na-fmoc-ne-boc-l-lysine, nepsilon-boc-nalpha-fmoc-l-lysine, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
C26H32N2O6
UMRUUWFGLGNQLI-QFIPXVFZSA-N
2724628
CC(C)(C)OC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Safety and Handling

Safety and Handling

EINECSNumber : 276-256-4

Recommended Storage : 4°C

SDS Documents
Product Certifications