missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-α-FMOC-N-epsilon-BOC-L-lysine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC265220010

 View more versions of this product

Catalog No. AC265220010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-α-FMOC-N-ε-BOC-L-lysine
Glass bottle
− 12.00
129.0°C to 135.0°C
71989-26-9
100.0
C26H32N2O6
a-boc-l-lysine
CC(C)(C)OC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
468.54
468.54
98%
Authentic
−12° (24°C c=2,DMF)
White
1g
97.5
98%
FMOC-NHCH(COOH)(CH2)4NHC(O)OC(CH3)3
UMRUUWFGLGNQLI-QFIPXVFZSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
2724628
Powder

Chemical Identifiers

71989-26-9
468.54
a-boc-l-lysine
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
C26H32N2O6
UMRUUWFGLGNQLI-QFIPXVFZSA-N
2724628
CC(C)(C)OC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Safety and Handling

Safety and Handling

Recommended Storage : 4°C

SDS Documents