missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-α-FMOC-N-epsilon-BOC-L-lysine, 98%, ACROS Organics™

Chemical Identifiers

CAS 71989-26-9
Molecular Formula C26H32N2O6
Molecular Weight (g/mol) 468.54
InChI Key UMRUUWFGLGNQLI-QFIPXVFZSA-N
Synonym a-boc-l-lysine, a-fmoc-n, fmoc-lys boc, fmoc-lys boc-oh, n, n-alpha-fmoc-n-epsilon-tert-boc-l-lysine, n-alpha-fmoc-nepsilon-boc-l-lysine, n-fmoc-n'-boc-l-lysine, na-fmoc-ne-boc-l-lysine, nepsilon-boc-nalpha-fmoc-l-lysine, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid
PubChem CID 2724628
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC265220010
SDS View Documents
Thermo Fisher Scientific
AC265220010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC265220100
SDS View Documents
Thermo Fisher Scientific
AC265220100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC265220250
SDS View Documents
Thermo Fisher Scientific
AC265220250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

71989-26-9
468.54
a-boc-l-lysine, a-fmoc-n, fmoc-lys boc, fmoc-lys boc-oh, n, n-alpha-fmoc-n-epsilon-tert-boc-l-lysine, n-alpha-fmoc-nepsilon-boc-l-lysine, n-fmoc-n'-boc-l-lysine, na-fmoc-ne-boc-l-lysine, nepsilon-boc-nalpha-fmoc-l-lysine, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
C26H32N2O6
UMRUUWFGLGNQLI-QFIPXVFZSA-N
2724628
CC(C)(C)OC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Authentic
−12° (24°C c=2,DMF)
White
1g
97.5
97.5% min. (HPLC)
FMOC-NHCH(COOH)(CH2)4NHC(O)OC(CH3)3
UMRUUWFGLGNQLI-QFIPXVFZSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
2724628
Powder
N-α-FMOC-N-ε-BOC-L-lysine
Glass bottle
− 12.00
129.0°C to 135.0°C
71989-26-9
100.0
C26H32N2O6
a-boc-l-lysine, a-fmoc-n, fmoc-lys boc, fmoc-lys boc-oh, n, n-alpha-fmoc-n-epsilon-tert-boc-l-lysine, n-alpha-fmoc-nepsilon-boc-l-lysine, n-fmoc-n'-boc-l-lysine, na-fmoc-ne-boc-l-lysine, nepsilon-boc-nalpha-fmoc-l-lysine, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid
CC(C)(C)OC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
468.54
468.54
98%
Safety and Handling

Safety and Handling

EINECSNumber : 276-256-4

Recommended Storage : 4°C

SDS