missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-α-FMOC-N-delta-Trityl-L-glutamine, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC440640250
Specifications
N-α-FMOC-N-δ-Trityl-L-glutamine | |
− 14.20 | |
131.0°C to 144.0°C | |
132327-80-1,75-09-2 | |
100.0 | |
C39H34N2O5 | |
WDGICUODAOGOMO-DHUJRADRSA-N | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoic acid | |
10919157 | |
Powder |
Authentic | |
White to Yellow | |
25g | |
94.0 | |
97.5% min. (HPLC) | |
2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, a-fmoc-n, a-trityl-l-glutamine, fmoc-gln trt-oh, fmoc-glutamine trt-oh, fmoc-l-gln trt-oh, ksc180c8t, n, n-fmoc-n5-trityl-l-glutamine, nalpha-fmoc-ndelta-trityl-l-glutamine, pubchem10017, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-oxo-5-tritylamino pentanoic acid | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46 | |
610.71 | |
610.71 | |
95% |
Chemical Identifiers
132327-80-1 | |
610.71 | |
2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-triphenylmethylcarbamoyl butanoic acid, a-fmoc-n, a-trityl-l-glutamine, fmoc-gln trt-oh, fmoc-glutamine trt-oh, fmoc-l-gln trt-oh, ksc180c8t, n, n-fmoc-n5-trityl-l-glutamine, nalpha-fmoc-ndelta-trityl-l-glutamine, pubchem10017, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-oxo-5-tritylamino pentanoic acid | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-(tritylamino)pentanoic acid |
C39H34N2O5 | |
WDGICUODAOGOMO-DHUJRADRSA-N | |
10919157 | |
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)NC(=O)CCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning