Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-valine pentafluorophenyl ester, 98%

Chemical Identifiers

Chemical Name or Material N-Fmoc-L-valine pentafluorophenyl ester
CAS 86060-87-9
Formula Weight 505.44
MDL Number MFCD00037650
Beilstein 3640481
Synonym Fmoc-Val-OPfp
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6681803

  SDS
alfa aesar™
H6681803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6681806

  SDS
alfa aesar™
H6681806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 505.44
Chemical Name or Material N-Fmoc-L-valine pentafluorophenyl ester
Physical Form Solid
MDL Number MFCD00037650
CAS 86060-87-9
Beilstein 3640481
Synonym Fmoc-Val-OPfp
Assay 98%
Quantity 1g