Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-tryptophan pentafluorophenyl ester, 98%

Chemical Identifiers

Chemical Name or Material N-Fmoc-L-tryptophan pentafluorophenyl ester
CAS 86069-87-6
Formula Weight 592.52
MDL Number MFCD00065681
Beilstein 4228691
Synonym Fmoc-Trp-OPfp
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6606903

 
alfa aesar™
H6606903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6606906

 
alfa aesar™
H6606906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 592.52
MDL Number MFCD00065681
CAS 86069-87-6
Synonym Fmoc-Trp-OPfp
Physical Form Powder
Beilstein 4228691
Color White
Quantity 1g
Storage Note 1 2° to 8°C
Chemical Name or Material N-Fmoc-L-tryptophan pentafluorophenyl ester
Assay 98%
Safety and Handling

2° to 8°C