missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-threoninol, 98%

$32.38 - $380.91

Chemical Identifiers

CAS 176380-53-3
Molecular Formula C19H21NO4
Molecular Weight (g/mol) 327.38
MDL Number MFCD00270218
InChI Key YOKDHMTZJSRRIQ-KZULUSFZSA-N
Synonym fmoc-threoninol, n-fmoc-l-threonol, fmoc-thr-ol, fmoc-l-threoninol, n-fmoc-l-threoninol, 9h-fluoren-9-yl methyl 2r,3r-1,3-dihydroxybutan-2-yl carbamate, 1r,2r-2-hydroxy-1-hydroxymethylpropyl carbamic acid 9h-fluoren-9-ylmethyl ester, 9-fluorenylmethyloxycarbonyl-l-threoninol, 9h-fluoren-9-ylmethyl n-2r,3r-1,3-dihydroxybutan-2-yl carbamate, fmoc-l-threonol
PubChem CID 7020321
IUPAC Name 9H-fluoren-9-ylmethyl N-[(2R,3R)-1,3-dihydroxybutan-2-yl]carbamate
SMILES CC(C(CO)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66211MD View Documents Alfa Aesar™
H66211MD
250mg Each for $32.38
 
AAH6621103 View Documents Alfa Aesar™
H6621103
1g Each for $95.46
 
AAH6621106 View Documents Alfa Aesar™
H6621106
5g Each for $380.91
 
Specifications

Chemical Identifiers

176380-53-3
327.38
YOKDHMTZJSRRIQ-KZULUSFZSA-N
7020321
CC(C(CO)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)O
C19H21NO4
MFCD00270218
fmoc-threoninol, n-fmoc-l-threonol, fmoc-thr-ol, fmoc-l-threoninol, n-fmoc-l-threoninol, 9h-fluoren-9-yl methyl 2r,3r-1,3-dihydroxybutan-2-yl carbamate, 1r,2r-2-hydroxy-1-hydroxymethylpropyl carbamic acid 9h-fluoren-9-ylmethyl ester, 9-fluorenylmethyloxycarbonyl-l-threoninol, 9h-fluoren-9-ylmethyl n-2r,3r-1,3-dihydroxybutan-2-yl carbamate, fmoc-l-threonol
9H-fluoren-9-ylmethyl N-[(2R,3R)-1,3-dihydroxybutan-2-yl]carbamate

Specifications

250mg
C19H21NO4
fmoc-threoninol, n-fmoc-l-threonol, fmoc-thr-ol, fmoc-l-threoninol, n-fmoc-l-threoninol, 9h-fluoren-9-yl methyl 2r,3r-1,3-dihydroxybutan-2-yl carbamate, 1r,2r-2-hydroxy-1-hydroxymethylpropyl carbamic acid 9h-fluoren-9-ylmethyl ester, 9-fluorenylmethyloxycarbonyl-l-threoninol, 9h-fluoren-9-ylmethyl n-2r,3r-1,3-dihydroxybutan-2-yl carbamate, fmoc-l-threonol
CC(C(CO)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)O
327.38
327.38
98%
176380-53-3
MFCD00270218
YOKDHMTZJSRRIQ-KZULUSFZSA-N
9H-fluoren-9-ylmethyl N-[(2R,3R)-1,3-dihydroxybutan-2-yl]carbamate
7020321
Solid
N-Fmoc-L-threoninol
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C