Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-threonine monohydrate, 98%

$43.08 - $43.08

Chemical Identifiers

Chemical Name or Material N-Fmoc-L-threonine monohydrate
CAS 73731-37-0
Molecular Formula C19H19NO5·H2O
Formula Weight 359.38 (341.36 Anhydrous)
MDL Number MFCD00077072
Synonym N-(9-Fluorenylmethoxycarbonyl)-L-threonine; Fmoc-Thr-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2112003

  SDS
alfa aesar™
B2112003
1g Each for $43.08
Description & Specifications

Specifications

Formula Weight 359.38 (341.36 Anhydrous)
Optical Rotation −16° (c=1 in DMF)
Assay 98%
Chemical Name or Material N-Fmoc-L-threonine monohydrate
MDL Number MFCD00077072
Quantity 1g
Molecular Formula C19H19NO5·H2O
Melting Point 94° to 96°C
Recommended Storage 2° to 8°C
Synonym N-(9-Fluorenylmethoxycarbonyl)-L-threonine; Fmoc-Thr-OH
CAS 73731-37-0
Safety and Handling

2° to 8°C