Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-threonine, 98%

Chemical Identifiers

Chemical Name or Material N-Fmoc-L-threonine
CAS 118609-38-4
Formula Weight 341.36
MDL Number MFCD00153365
Synonym Fmoc-D-Thr-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6660506

  SDS
alfa aesar™
H6660506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6660503

  SDS
alfa aesar™
H6660503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 341.36
Assay 98%
MDL Number MFCD00153365
CAS 118609-38-4
Synonym Fmoc-D-Thr-OH
Physical Form Solid
Storage Note 1 2° to 8°C
Chemical Name or Material N-Fmoc-L-threonine
Quantity 5g

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Sparingly soluble in water (0.053 g/L) (25°C).

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

2° to 8°C