missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-L-threonine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6660506
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is employed as a intermediate for pharmaceutical.
Solubility
Sparingly soluble in water (0.053 g/L) (25°C).
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
N-Fmoc-L-threonine | |
C19H19NO5 | |
2r,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-hydroxybutanoic acid hydrate, 2r,3s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxybutanoic acid hydrate, fmoc-d-thr-oh.h2o, n-alpha-9-fluorenylmethyloxycarbonyl-d-threonine monohydrate, n-9h-fluoren-9-yl methoxy carbonyl-d-threonine-water 1/1 | |
OYULCCKKLJPNPU-DIFFPNOSSA-N | |
(2S,3R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxybutanoic acid | |
71607443 | |
98% | |
Solid |
118609-38-4 | |
MFCD00077072 | |
Sparingly soluble in water (0.053g/L) (25°C). | |
C[C@@H](O)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O | |
341.36 | |
341.36 | |
5 g |
Chemical Identifiers
118609-38-4 | |
341.36 | |
OYULCCKKLJPNPU-DIFFPNOSSA-N | |
71607443 | |
C[C@@H](O)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O |
C19H19NO5 | |
MFCD00077072 | |
2r,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-hydroxybutanoic acid hydrate, 2r,3s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxybutanoic acid hydrate, fmoc-d-thr-oh.h2o, n-alpha-9-fluorenylmethyloxycarbonyl-d-threonine monohydrate, n-9h-fluoren-9-yl methoxy carbonyl-d-threonine-water 1/1 | |
(2S,3R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-hydroxybutanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold