missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-prolyl-L-proline, 95%

$121.51 - $453.38

Chemical Identifiers

CAS 129223-22-9
Molecular Formula C25H26N2O5
Molecular Weight (g/mol) 434.492
MDL Number MFCD00237673
InChI Key VRAQFWSWKRNOGU-VXKWHMMOSA-N
Synonym fmoc-pro-pro-oh, s-1-s-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic acid, l-proline, 1-9h-fluoren-9-ylmethoxy carbonyl-l-prolyl, 2s-1-2s-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic acid, s-1-s-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic aci, fmoc-pro-pro, fmoc-prolinyl-proline, fmoc-prolylproline-oh, fmoc-l-pro-l-pro-oh, fmoc-prolinyl-proline s,s
PubChem CID 11133845
IUPAC Name (2S)-1-[(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)pyrrolidine-2-carbonyl]pyrrolidine-2-carboxylic acid
SMILES C1CC(N(C1)C(=O)C2CCCN2C(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6622803 View Documents Alfa Aesar™
H6622803
1g Each for $121.51
 
AAH6622806 View Documents Alfa Aesar™
H6622806
5g Each for $453.38
 
Specifications

Chemical Identifiers

129223-22-9
434.492
VRAQFWSWKRNOGU-VXKWHMMOSA-N
11133845
C1CC(N(C1)C(=O)C2CCCN2C(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)C(=O)O
C25H26N2O5
MFCD00237673
fmoc-pro-pro-oh, s-1-s-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic acid, l-proline, 1-9h-fluoren-9-ylmethoxy carbonyl-l-prolyl, 2s-1-2s-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic acid, s-1-s-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic aci, fmoc-pro-pro, fmoc-prolinyl-proline, fmoc-prolylproline-oh, fmoc-l-pro-l-pro-oh, fmoc-prolinyl-proline s,s
(2S)-1-[(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)pyrrolidine-2-carbonyl]pyrrolidine-2-carboxylic acid

Specifications

129223-22-9
MFCD00237673
VRAQFWSWKRNOGU-VXKWHMMOSA-N
(2S)-1-[(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)pyrrolidine-2-carbonyl]pyrrolidine-2-carboxylic acid
11133845
Solid
1g
C25H26N2O5
fmoc-pro-pro-oh, s-1-s-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic acid, l-proline, 1-9h-fluoren-9-ylmethoxy carbonyl-l-prolyl, 2s-1-2s-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic acid, s-1-s-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidine-2-carbonyl pyrrolidine-2-carboxylic aci, fmoc-pro-pro, fmoc-prolinyl-proline, fmoc-prolylproline-oh, fmoc-l-pro-l-pro-oh, fmoc-prolinyl-proline s,s
C1CC(N(C1)C(=O)C2CCCN2C(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)C(=O)O
434.492
434.49
95%
N-Fmoc-L-prolyl-L-proline
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C