missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-proline pentafluorophenyl ester, 98%

Manufacturer:  Alfa Aesar™ H6345703

 View more versions of this product

Catalog No. AAH6345703



Specifications

Specifications

N-Fmoc-L-proline pentafluorophenyl ester
C26H18F5NO4
4731096
CQBLOHXKGUNWRV-SFHVURJKSA-N
1-O-(9H-fluoren-9-ylmethyl) 2-O-(2,3,4,5,6-pentafluorophenyl) (2S)-pyrrolidine-1,2-dicarboxylate
2733412
Solid
127°C to 129°C
86060-90-4
MFCD00065672
fmoc-pro-opfp, fmoc-l-pro-opfp, fmoc-l-proline pentafluorophenyl ester, fmoc-pro-opfp, 1,2-pyrrolidinedicarboxylicacid, 1-9h-fluoren-9-ylmethyl 2-pentafluorophenyl ester, 2s, n-9-fluorenylmethoxycarbonyl-l-proline pentafluorophenyl ester, 1-9h-fluorene-9-ylmethoxycarbonyl-l-proline pentafluorophenyl ester, 1-9h-fluoren-9-ylmethyl 2-2,3,4,5,6-pentafluorophenyl 2s-pyrrolidine-1,2-dicarboxylate, 1-o-9h-fluoren-9-ylmethyl 2-o-2,3,4,5,6-pentafluorophenyl 2s-pyrrolidine-1,2-dicarboxylate
C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)OC5=C(C(=C(C(=C5F)F)F)F)F
503.425
503.42
98%
1g

Chemical Identifiers

86060-90-4
503.425
CQBLOHXKGUNWRV-SFHVURJKSA-N
2733412
C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)OC5=C(C(=C(C(=C5F)F)F)F)F
C26H18F5NO4
MFCD00065672
fmoc-pro-opfp, fmoc-l-pro-opfp, fmoc-l-proline pentafluorophenyl ester, fmoc-pro-opfp, 1,2-pyrrolidinedicarboxylicacid, 1-9h-fluoren-9-ylmethyl 2-pentafluorophenyl ester, 2s, n-9-fluorenylmethoxycarbonyl-l-proline pentafluorophenyl ester, 1-9h-fluorene-9-ylmethoxycarbonyl-l-proline pentafluorophenyl ester, 1-9h-fluoren-9-ylmethyl 2-2,3,4,5,6-pentafluorophenyl 2s-pyrrolidine-1,2-dicarboxylate, 1-o-9h-fluoren-9-ylmethyl 2-o-2,3,4,5,6-pentafluorophenyl 2s-pyrrolidine-1,2-dicarboxylate
1-O-(9H-fluoren-9-ylmethyl) 2-O-(2,3,4,5,6-pentafluorophenyl) (2S)-pyrrolidine-1,2-dicarboxylate
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

Documents