Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-proline N-succinimidyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Fmoc-L-proline N-succinimidyl ester
CAS 109074-94-4
Formula Weight 434.45
MDL Number MFCD00065673
Synonym Fmoc-Pro-OSu; N-Fmoc-L-proline N-hydroxysuccinimide ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6650706

 
alfa aesar™
H6650706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6650714

 
alfa aesar™
H6650714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 434.45
Chemical Name or Material N-Fmoc-L-proline N-succinimidyl ester
MDL Number MFCD00065673
Assay 95%
Physical Form Solid
Quantity 5g
CAS 109074-94-4
Storage Note 1 -30° to -10°C
Synonym Fmoc-Pro-OSu; N-Fmoc-L-proline N-hydroxysuccinimide ester
Safety and Handling

-30° to -10°C