missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-phenylalaninol, 95%

$165.01 - $652.31

Chemical Identifiers

CAS 129397-83-7
Molecular Formula C24H23NO3
Molecular Weight (g/mol) 373.452
MDL Number MFCD00235941
InChI Key SJGBJASOHDROCR-SFHVURJKSA-N
Synonym fmoc-phenylalaninol, fmoc-l-phenylalaninol, s-9h-fluoren-9-yl methyl 1-hydroxy-3-phenylpropan-2-yl carbamate, fmoc-phe-ol, s-9h-fluoren-9-yl methyl 1-hydroxy-3-phenylpropan-2-ylcarbamate, n-fmoc-l-phenylalaninol, s-2-9-fluorenylmethyloxycarbonyl amino-3-phenyl-1-propanol, 9h-fluoren-9-ylmethyl n-2s-1-hydroxy-3-phenylpropan-2-yl carbamate, pubchem12466, carbamic acid,n-1s-1-hydroxymethyl-2-phenylethyl-, 9h-fluoren-9-ylmethyl ester
PubChem CID 7019531
IUPAC Name 9H-fluoren-9-ylmethyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate
SMILES C1=CC=C(C=C1)CC(CO)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6638106 View Documents Alfa Aesar™
H6638106
5g Each for $165.01
 
AAH6638114 View Documents Alfa Aesar™
H6638114
25g Each for $652.31
 
Specifications

Chemical Identifiers

129397-83-7
373.452
SJGBJASOHDROCR-SFHVURJKSA-N
7019531
C1=CC=C(C=C1)CC(CO)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C24H23NO3
MFCD00235941
fmoc-phenylalaninol, fmoc-l-phenylalaninol, s-9h-fluoren-9-yl methyl 1-hydroxy-3-phenylpropan-2-yl carbamate, fmoc-phe-ol, s-9h-fluoren-9-yl methyl 1-hydroxy-3-phenylpropan-2-ylcarbamate, n-fmoc-l-phenylalaninol, s-2-9-fluorenylmethyloxycarbonyl amino-3-phenyl-1-propanol, 9h-fluoren-9-ylmethyl n-2s-1-hydroxy-3-phenylpropan-2-yl carbamate, pubchem12466, carbamic acid,n-1s-1-hydroxymethyl-2-phenylethyl-, 9h-fluoren-9-ylmethyl ester
9H-fluoren-9-ylmethyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate

Specifications

5g
C24H23NO3
fmoc-phenylalaninol, fmoc-l-phenylalaninol, s-9h-fluoren-9-yl methyl 1-hydroxy-3-phenylpropan-2-yl carbamate, fmoc-phe-ol, s-9h-fluoren-9-yl methyl 1-hydroxy-3-phenylpropan-2-ylcarbamate, n-fmoc-l-phenylalaninol, s-2-9-fluorenylmethyloxycarbonyl amino-3-phenyl-1-propanol, 9h-fluoren-9-ylmethyl n-2s-1-hydroxy-3-phenylpropan-2-yl carbamate, pubchem12466, carbamic acid,n-1s-1-hydroxymethyl-2-phenylethyl-, 9h-fluoren-9-ylmethyl ester
C1=CC=C(C=C1)CC(CO)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
373.452
373.45
95%
129397-83-7
MFCD00235941
SJGBJASOHDROCR-SFHVURJKSA-N
9H-fluoren-9-ylmethyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate
7019531
Solid
N-Fmoc-L-phenylalaninol
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C