missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-phenylalanine pentafluorophenyl ester, 96%

$57.77 - $589.27

Chemical Identifiers

CAS 86060-92-6
Molecular Formula C30H20F5NO4
Molecular Weight (g/mol) 553.485
MDL Number MFCD00065669
InChI Key WKHPSOMXNCTXPK-QFIPXVFZSA-N
Synonym 2,3,4,5,6-pentafluorophenyl 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-phenylpropanoate, fmoc-l-phenylalanine pentafluorophenyl ester, fmoc-phe-opfp, l-phenylalanine,n-9h-fluoren-9-ylmethoxy carbonyl-, 2,3,4,5,6-pentafluorophenyl ester, n-9-fluorenylmethoxycarbonyl-l-phenylalanine pentafluorophenyl ester, n-9h-fluorene-9-ylmethoxycarbonyl-l-phenylalanine pentafluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-phenylalanine pentafluorphenyl ester, n-fmoc-l-phenylalanine pentafluorophenyl ester, pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-l-phenylalaninate
PubChem CID 2733280
IUPAC Name (2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoate
SMILES C1=CC=C(C=C1)CC(C(=O)OC2=C(C(=C(C(=C2F)F)F)F)F)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65319MD View Documents Alfa Aesar™
H65319MD
250mg Each for $57.77
 
AAH6531903 View Documents Alfa Aesar™
H6531903
1g Each for $141.82
 
AAH6531906 View Documents Alfa Aesar™
H6531906
5g Each for $589.27
 
Specifications

Chemical Identifiers

86060-92-6
553.485
WKHPSOMXNCTXPK-QFIPXVFZSA-N
2733280
C1=CC=C(C=C1)CC(C(=O)OC2=C(C(=C(C(=C2F)F)F)F)F)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
C30H20F5NO4
MFCD00065669
2,3,4,5,6-pentafluorophenyl 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-phenylpropanoate, fmoc-l-phenylalanine pentafluorophenyl ester, fmoc-phe-opfp, l-phenylalanine,n-9h-fluoren-9-ylmethoxy carbonyl-, 2,3,4,5,6-pentafluorophenyl ester, n-9-fluorenylmethoxycarbonyl-l-phenylalanine pentafluorophenyl ester, n-9h-fluorene-9-ylmethoxycarbonyl-l-phenylalanine pentafluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-phenylalanine pentafluorphenyl ester, n-fmoc-l-phenylalanine pentafluorophenyl ester, pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-l-phenylalaninate
(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoate

Specifications

86060-92-6
MFCD00065669
2,3,4,5,6-pentafluorophenyl 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-phenylpropanoate, fmoc-l-phenylalanine pentafluorophenyl ester, fmoc-phe-opfp, l-phenylalanine,n-9h-fluoren-9-ylmethoxy carbonyl-, 2,3,4,5,6-pentafluorophenyl ester, n-9-fluorenylmethoxycarbonyl-l-phenylalanine pentafluorophenyl ester, n-9h-fluorene-9-ylmethoxycarbonyl-l-phenylalanine pentafluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-phenylalanine pentafluorphenyl ester, n-fmoc-l-phenylalanine pentafluorophenyl ester, pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-l-phenylalaninate
C1=CC=C(C=C1)CC(C(=O)OC2=C(C(=C(C(=C2F)F)F)F)F)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35
553.485
553.49
96%
149°C to 153°C
N-Fmoc-L-phenylalanine pentafluorophenyl ester
C30H20F5NO4
4241247
WKHPSOMXNCTXPK-QFIPXVFZSA-N
(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoate
2733280
Powder
White
250mg
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold