Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-phenylalanine, 98+%

$25.54 - $83.27

Chemical Identifiers

Chemical Name or Material N-Fmoc-L-phenylalanine
CAS 35661-40-6
Molecular Formula C24H21NO4
Formula Weight 387.44
MDL Number MFCD00037128
Beilstein 3597808
Synonym N-(9-Fluorenylmethoxycarbonyl)-L-phenylalanine; Fmoc-Phe-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2121006

 
alfa aesar™
B2121006
5g Each for $25.54

AAB2121014

 
alfa aesar™
B2121014
25g Each for $83.27
Description & Specifications

Specifications

Formula Weight 387.44
Chemical Name or Material N-Fmoc-L-phenylalanine
Assay ≥98%
Molecular Formula C24H21NO4
Melting Point 180° to 185°C
MDL Number MFCD00037128
CAS 35661-40-6
Beilstein 3597808
Quantity 5g
Synonym N-(9-Fluorenylmethoxycarbonyl)-L-phenylalanine; Fmoc-Phe-OH
Optical Rotation −40° (c=1 in DMF)
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C