Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-methionine sulfone, 98%

Chemical Identifiers

Chemical Name or Material N-Fmoc-L-methionine sulfone
CAS 163437-14-7
Formula Weight 403.45
MDL Number MFCD00153361
Synonym Fmoc-Met(O2)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6303303

 
alfa aesar™
H6303303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6303306

 
alfa aesar™
H6303306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63033MD

 
alfa aesar™
H63033MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 403.45
Chemical Name or Material N-Fmoc-L-methionine sulfone
Physical Form Powder
Color White
Quantity 1g
Assay 98%
MDL Number MFCD00153361
CAS 163437-14-7
Synonym Fmoc-Met(O2)-OH