missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-L-methionine sulfone, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6303303
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-Fmoc-L-methionine sulfone | |
White | |
MFCD00153361 | |
KJLKPACOHZKRFM-SFHVURJKSA-N | |
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methanesulfonylbutanoic acid | |
7016362 | |
98% | |
Powder |
163437-14-7 | |
C20H21NO6S | |
fmoc-met o2-oh, fmoc-l-methionine sulfone, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylsulfonyl butanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methanesulfonylbutanoic acid, ambotzfaa1406, fmoc-met o 2-oh, pubchem22746, n-alpha-9-fluorenylmethyloxycarbonyl-l-methioninesulfon, s-2-9h-fluoren-9-yl methoxy carbonyl-amino-4-methylsulfonyl butanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino-4-methylsulfonyl butanoic acid | |
CS(=O)(=O)CC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
403.45 | |
403.45 | |
1 g |
Chemical Identifiers
163437-14-7 | |
403.45 | |
KJLKPACOHZKRFM-SFHVURJKSA-N | |
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-methanesulfonylbutanoic acid |
C20H21NO6S | |
MFCD00153361 | |
7016362 | |
CS(=O)(=O)CC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures