Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-methionine, 98+%

$25.72 - $102.82

Chemical Identifiers

Chemical Name or Material N-Fmoc-L-methionine
CAS 71989-28-1
Molecular Formula C20H21NO4S
Formula Weight 371.46
MDL Number MFCD00037134
Beilstein 4300266
Synonym N-(9-Fluorenylmethoxycarbonyl)-L-methionine; Fmoc-Met-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2122006

 
alfa aesar™
B2122006
5g Each for $25.72

AAB2122014

 
alfa aesar™
B2122014
25g Each for $102.82
Description & Specifications

Specifications

Formula Weight 371.46
Chemical Name or Material N-Fmoc-L-methionine
Assay ≥98%
Molecular Formula C20H21NO4S
Melting Point 132° to 135°C
MDL Number MFCD00037134
CAS 71989-28-1
Beilstein 4300266
Quantity 5g
Synonym N-(9-Fluorenylmethoxycarbonyl)-L-methionine; Fmoc-Met-OH
Optical Rotation −30° (c=1 in DMF)
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C