missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-leucine pentafluorophenyl ester, 98%

$32.53 - $118.43

Chemical Identifiers

CAS 86060-88-0
Molecular Formula C27H22F5NO4
Molecular Weight (g/mol) 519.468
MDL Number MFCD00065658
InChI Key NTQJCLLWLHKJLU-IBGZPJMESA-N
Synonym fmoc-leu-opfp, fmoc-l-leucine pentafluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-leucine pentafluorphenyl ester, 2,3,4,5,6-pentafluorophenyl 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylpentanoate, fmoc-leu-opfp, pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-l-leucinate, n-9h-fluorene-9-ylmethoxycarbonyl-l-leucine pentafluorophenyl ester, pentafluorophenyl n-9h-fluoren-9-yl methoxy carbonyl-l-leucinate
PubChem CID 13585944
IUPAC Name (2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoate
SMILES CC(C)CC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6382003 View Documents Alfa Aesar™
H6382003
1g Each for $32.53
 
AAH6382006 View Documents Alfa Aesar™
H6382006
5g Each for $118.43
 
Specifications

Chemical Identifiers

86060-88-0
519.468
NTQJCLLWLHKJLU-IBGZPJMESA-N
13585944
CC(C)CC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C27H22F5NO4
MFCD00065658
fmoc-leu-opfp, fmoc-l-leucine pentafluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-leucine pentafluorphenyl ester, 2,3,4,5,6-pentafluorophenyl 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylpentanoate, fmoc-leu-opfp, pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-l-leucinate, n-9h-fluorene-9-ylmethoxycarbonyl-l-leucine pentafluorophenyl ester, pentafluorophenyl n-9h-fluoren-9-yl methoxy carbonyl-l-leucinate
(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoate

Specifications

86060-88-0
MFCD00065658
fmoc-leu-opfp, fmoc-l-leucine pentafluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-leucine pentafluorphenyl ester, 2,3,4,5,6-pentafluorophenyl 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylpentanoate, fmoc-leu-opfp, pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-l-leucinate, n-9h-fluorene-9-ylmethoxycarbonyl-l-leucine pentafluorophenyl ester, pentafluorophenyl n-9h-fluoren-9-yl methoxy carbonyl-l-leucinate
CC(C)CC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
519.468
519.46
98%
1g
C27H22F5NO4
4731726
NTQJCLLWLHKJLU-IBGZPJMESA-N
(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoate
13585944
Powder
White
N-Fmoc-L-leucine pentafluorophenyl ester
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C