missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-leucine pentafluorophenyl ester, 98%

Manufacturer:  Alfa Aesar™ H6382006

 View more versions of this product

Catalog No. AAH6382006



Specifications

Specifications

N-Fmoc-L-leucine pentafluorophenyl ester
C27H22F5NO4
4731726
NTQJCLLWLHKJLU-IBGZPJMESA-N
(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoate
13585944
Powder
White
86060-88-0
MFCD00065658
fmoc-leu-opfp, fmoc-l-leucine pentafluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-leucine pentafluorphenyl ester, 2,3,4,5,6-pentafluorophenyl 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylpentanoate, fmoc-leu-opfp, pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-l-leucinate, n-9h-fluorene-9-ylmethoxycarbonyl-l-leucine pentafluorophenyl ester, pentafluorophenyl n-9h-fluoren-9-yl methoxy carbonyl-l-leucinate
CC(C)CC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
519.468
519.46
98%
5g

Chemical Identifiers

86060-88-0
519.468
NTQJCLLWLHKJLU-IBGZPJMESA-N
13585944
CC(C)CC(C(=O)OC1=C(C(=C(C(=C1F)F)F)F)F)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C27H22F5NO4
MFCD00065658
fmoc-leu-opfp, fmoc-l-leucine pentafluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-leucine pentafluorphenyl ester, 2,3,4,5,6-pentafluorophenyl 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylpentanoate, fmoc-leu-opfp, pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-l-leucinate, n-9h-fluorene-9-ylmethoxycarbonyl-l-leucine pentafluorophenyl ester, pentafluorophenyl n-9h-fluoren-9-yl methoxy carbonyl-l-leucinate
(2,3,4,5,6-pentafluorophenyl) (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoate
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents