missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-L-isoleucine, 98%, Thermo Scientific™
$36.40 - $36.40
Chemical Identifiers
CAS | 71989-23-6 |
---|---|
Molecular Formula | C21H23NO4 |
Molecular Weight (g/mol) | 353.42 |
MDL Number | MFCD00037125 |
InChI Key | QXVFEIPAZSXRGM-DJJJIMSYSA-N |
Synonym | fmoc-ile-oh, fmoc-l-isoleucine, n-fmoc-l-isoleucine, fmoc-l-ile-oh, n-9h-fluoren-9-ylmethoxy carbonyl-l-isoleucine, n-9-fluorenylmethoxycarbonyl-l-isoleucine, fmoc-ile, fmoc-l-ile, 2s,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylpentanoic acid, 9-fluorenylmethoxycarbonyl-l-isoleucine |
PubChem CID | 2724629 |
SMILES | CC[C@H](C)[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2115403
|
Thermo Scientific Chemicals
B2115403 |
1 g |
Each for $36.40
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsN-Fmoc-L-isoleucine is an unnatural amino acid used for peptidomimetics. It is also used as a pharmaceutical intermediate.
Solubility
Soluble in methanol (very faint turbidity).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
71989-23-6 | |
353.42 | |
QXVFEIPAZSXRGM-DJJJIMSYSA-N | |
2724629 |
C21H23NO4 | |
MFCD00037125 | |
fmoc-ile-oh, fmoc-l-isoleucine, n-fmoc-l-isoleucine, fmoc-l-ile-oh, n-9h-fluoren-9-ylmethoxy carbonyl-l-isoleucine, n-9-fluorenylmethoxycarbonyl-l-isoleucine, fmoc-ile, fmoc-l-ile, 2s,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylpentanoic acid, 9-fluorenylmethoxycarbonyl-l-isoleucine | |
CC[C@H](C)[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Specifications
71989-23-6 | |
C21H23NO4 | |
1 g | |
fmoc-ile-oh, fmoc-l-isoleucine, n-fmoc-l-isoleucine, fmoc-l-ile-oh, n-9h-fluoren-9-ylmethoxy carbonyl-l-isoleucine, n-9-fluorenylmethoxycarbonyl-l-isoleucine, fmoc-ile, fmoc-l-ile, 2s,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylpentanoic acid, 9-fluorenylmethoxycarbonyl-l-isoleucine | |
QXVFEIPAZSXRGM-DJJJIMSYSA-N | |
(2S,3S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-methylpentanoic acid | |
2724629 | |
98% |
146°C to 148°C | |
MFCD00037125 | |
4716717 | |
Soluble in methanol (very faint turbidity). | |
CC[C@H](C)[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
353.42 | |
353.42 | |
N-Fmoc-L-isoleucine |
Safety and Handling
EINECSNumber : 276-255-9
TSCA : No
Recommended Storage : Keep cold