Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-homoarginine, 96%

Chemical Identifiers

Chemical Name or Material N-Fmoc-L-homoarginine
CAS 776277-76-0
Formula Weight 410.47
MDL Number MFCD01861360
Synonym Fmoc-HoArg-OH; (S)-(2-Fmoc-amino)-6-guanidinohexanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6322803

  SDS
alfa aesar™
H6322803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6322806

  SDS
alfa aesar™
H6322806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63228MD

  SDS
alfa aesar™
H63228MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 410.47
Chemical Name or Material N-Fmoc-L-homoarginine
Physical Form Powder
Assay 96%
MDL Number MFCD01861360
CAS 776277-76-0
Synonym Fmoc-HoArg-OH; (S)-(2-Fmoc-amino)-6-guanidinohexanoic acid
Color White
Quantity 1g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C