missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-homoarginine, 96%

Manufacturer:  Alfa Aesar™ H63228MD

 View more versions of this product

Catalog No. AAH63228MD



Specifications

Specifications

N-Fmoc-L-homoarginine
C22H26N4O4
fmoc-hoarg-oh, fmoc-homoarg-oh, fmoc-har-oh, fmoc-l-homoarginine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-guanidinohexanoic acid, fmoc-homoarg, 2s-6-carbamimidamido-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, n~6~-diaminomethylidene-n~2~-9h-fluoren-9-yl methoxy carbonyl-l-lysine, 2s-6-diaminomethylidene amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCCN=C(N)N)C(=O)O
410.474
410.47
96%
250mg
776277-76-0
MFCD01861360
QRELIJBJYZHTQU-IBGZPJMESA-N
(2S)-6-(diaminomethylideneamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
40418724
Powder
White

Chemical Identifiers

776277-76-0
410.474
QRELIJBJYZHTQU-IBGZPJMESA-N
40418724
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCCN=C(N)N)C(=O)O
C22H26N4O4
MFCD01861360
fmoc-hoarg-oh, fmoc-homoarg-oh, fmoc-har-oh, fmoc-l-homoarginine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-guanidinohexanoic acid, fmoc-homoarg, 2s-6-carbamimidamido-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, n~6~-diaminomethylidene-n~2~-9h-fluoren-9-yl methoxy carbonyl-l-lysine, 2s-6-diaminomethylidene amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid
(2S)-6-(diaminomethylideneamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents