missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-homoarginine, 96%

Manufacturer:  Alfa Aesar™ H6322806

 View more versions of this product

Catalog No. AAH6322806


 

Specifications

Specifications

N-Fmoc-L-homoarginine
96%
MFCD01861360
QRELIJBJYZHTQU-IBGZPJMESA-N
(2S)-6-(diaminomethylideneamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
40418724
Powder
5g
776277-76-0
C22H26N4O4
Fmoc-HoArg-OH; (S)-(2-Fmoc-amino)-6-guanidinohexanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCCN=C(N)N)C(=O)O
410.474
410.47
White

Chemical Identifiers

776277-76-0
410.474
QRELIJBJYZHTQU-IBGZPJMESA-N
40418724
C22H26N4O4
MFCD01861360
Fmoc-HoArg-OH; (S)-(2-Fmoc-amino)-6-guanidinohexanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents