missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-glutamic acid 5-cyclohexyl ester, 98%

$108.86 - $433.34

Chemical Identifiers

CAS 150047-85-1
Molecular Formula C26H29NO6
Molecular Weight (g/mol) 451.519
MDL Number MFCD00270538
InChI Key ODIPEUXZAYGDMU-QHCPKHFHSA-N
Synonym fmoc-glu ochx-oh, fmoc-glu ochex-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-cyclohexyloxy-5-oxopentanoic acid, ambotzfaa1719, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-cyclohexyl ester, 2s-5-cyclohexyloxy-2-9h-fluoren-9-yl methoxy carbonyl amino-5-oxopentanoic acid non-preferred name, 2s-5-cyclohexyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid
PubChem CID 51340555
IUPAC Name (2S)-5-cyclohexyloxy-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxopentanoic acid
SMILES C1CCC(CC1)OC(=O)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6614606 View Documents Alfa Aesar™
H6614606
5g Each for $108.86
 
AAH6614614 View Documents Alfa Aesar™
H6614614
25g Each for $433.34
 
Specifications

Chemical Identifiers

150047-85-1
451.519
ODIPEUXZAYGDMU-QHCPKHFHSA-N
51340555
C1CCC(CC1)OC(=O)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C26H29NO6
MFCD00270538
fmoc-glu ochx-oh, fmoc-glu ochex-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-cyclohexyloxy-5-oxopentanoic acid, ambotzfaa1719, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-cyclohexyl ester, 2s-5-cyclohexyloxy-2-9h-fluoren-9-yl methoxy carbonyl amino-5-oxopentanoic acid non-preferred name, 2s-5-cyclohexyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid
(2S)-5-cyclohexyloxy-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxopentanoic acid

Specifications

150047-85-1
MFCD00270538
ODIPEUXZAYGDMU-QHCPKHFHSA-N
(2S)-5-cyclohexyloxy-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxopentanoic acid
51340555
Solid
5g
C26H29NO6
fmoc-glu ochx-oh, fmoc-glu ochex-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-cyclohexyloxy-5-oxopentanoic acid, ambotzfaa1719, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-cyclohexyl ester, 2s-5-cyclohexyloxy-2-9h-fluoren-9-yl methoxy carbonyl amino-5-oxopentanoic acid non-preferred name, 2s-5-cyclohexyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid
C1CCC(CC1)OC(=O)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
451.519
451.52
98%
N-Fmoc-L-glutamic acid 5-cyclohexyl ester
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C