missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-glutamic acid 1-tert-butyl ester, 98%

$56.28 - $664.65

Chemical Identifiers

CAS 84793-07-7
Molecular Formula C24H27NO6
Molecular Weight (g/mol) 425.481
MDL Number MFCD00065648
InChI Key GOPWHXPXSPIIQZ-FQEVSTJZSA-N
Synonym fmoc-glu-otbu, fmoc-l-glutamic acid 1-tert-butyl ester, fmoc-glu obut-oh, fmoc-glu-obut, 4s-5-tert-butoxy-4-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, fmoc-glu-ot-bu, fmoc-glu-o-t-bu, pubchem10014, ksc448a0b
PubChem CID 7017912
IUPAC Name (4S)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)C(CCC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62146MD View Documents Alfa Aesar™
H62146MD
250mg Each for $56.28
 
AAH6214603 View Documents Alfa Aesar™
H6214603
1g Each for $168.09
 
AAH6214606 View Documents Alfa Aesar™
H6214606
5g Each for $664.65
 
Specifications

Chemical Identifiers

84793-07-7
425.481
GOPWHXPXSPIIQZ-FQEVSTJZSA-N
7017912
CC(C)(C)OC(=O)C(CCC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C24H27NO6
MFCD00065648
fmoc-glu-otbu, fmoc-l-glutamic acid 1-tert-butyl ester, fmoc-glu obut-oh, fmoc-glu-obut, 4s-5-tert-butoxy-4-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, fmoc-glu-ot-bu, fmoc-glu-o-t-bu, pubchem10014, ksc448a0b
(4S)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid

Specifications

250mg
C24H27NO6
fmoc-glu-otbu, fmoc-l-glutamic acid 1-tert-butyl ester, fmoc-glu obut-oh, fmoc-glu-obut, 4s-5-tert-butoxy-4-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, fmoc-glu-ot-bu, fmoc-glu-o-t-bu, pubchem10014, ksc448a0b
CC(C)(C)OC(=O)C(CCC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
425.481
425.47
N-Fmoc-L-glutamic acid 1-tert-butyl ester
84793-07-7
MFCD00065648
GOPWHXPXSPIIQZ-FQEVSTJZSA-N
(4S)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
7017912
98%