Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-glutamic acid 1-benzyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Fmoc-L-glutamic acid 1-benzyl ester
CAS 122350-52-1
Formula Weight 459.5
MDL Number MFCD00080272
Synonym Fmoc-Glu-OBzl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6655103

 
alfa aesar™
H6655103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6655106

 
alfa aesar™
H6655106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 459.5
Chemical Name or Material N-Fmoc-L-glutamic acid 1-benzyl ester
CAS 122350-52-1
Physical Form Powder
MDL Number MFCD00080272
Synonym Fmoc-Glu-OBzl
Storage Note 1 -30° to -10°C
Assay 95%
Color White
Quantity 1g
Safety and Handling

-30° to -10°C