Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-citrulline, 98%

Chemical Identifiers

Chemical Name or Material N-Fmoc-L-citrulline
CAS 133174-15-9
Formula Weight 397.42
MDL Number MFCD00151943
Beilstein 7780987
Synonym Fmoc-Cit-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6334203

 
alfa aesar™
H6334203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6334206

 
alfa aesar™
H6334206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6334214

 
alfa aesar™
H6334214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 397.42
Chemical Name or Material N-Fmoc-L-citrulline
Physical Form Solid
Assay 98%
MDL Number MFCD00151943
CAS 133174-15-9
Beilstein 7780987
Synonym Fmoc-Cit-OH
Storage Note 1 2° to 8°C
Quantity 1g
Safety and Handling

2° to 8°C