missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-aspartic acid 4-methylpentyl ester, 98%

$28.22 - $374.73

Chemical Identifiers

CAS 180675-08-5
Molecular Formula C25H29NO6
Molecular Weight (g/mol) 439.508
MDL Number MFCD10001333
InChI Key SYGKUYKLHYQKPL-NRFANRHFSA-N
Synonym fmoc-asp ompe-oh, l-aspartic acid,n-9h-fluoren-9-ylmethoxy carbonyl-, 4-1-ethyl-1-methylpropyl ester, n-9h-fluoren-9-ylmethoxycarbonyl-l-aspartic acid 4-1-methyl-1-ethylpropyl ester, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-3-methylpentan-3-yl oxy-4-oxobutanoic acid non-preferred name, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-3-methylpentan-3-yl oxy-4-oxobutanoic acid
PubChem CID 56777270
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-(3-methylpentan-3-yloxy)-4-oxobutanoic acid
SMILES CCC(C)(CC)OC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66281MD View Documents Alfa Aesar™
H66281MD
250mg Each for $28.22
 
AAH6628103 View Documents Alfa Aesar™
H6628103
1g Each for $90.07
 
AAH6628106 View Documents Alfa Aesar™
H6628106
5g Each for $374.73
 
Specifications

Chemical Identifiers

180675-08-5
439.508
SYGKUYKLHYQKPL-NRFANRHFSA-N
56777270
CCC(C)(CC)OC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C25H29NO6
MFCD10001333
fmoc-asp ompe-oh, l-aspartic acid,n-9h-fluoren-9-ylmethoxy carbonyl-, 4-1-ethyl-1-methylpropyl ester, n-9h-fluoren-9-ylmethoxycarbonyl-l-aspartic acid 4-1-methyl-1-ethylpropyl ester, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-3-methylpentan-3-yl oxy-4-oxobutanoic acid non-preferred name, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-3-methylpentan-3-yl oxy-4-oxobutanoic acid
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-(3-methylpentan-3-yloxy)-4-oxobutanoic acid

Specifications

180675-08-5
MFCD10001333
SYGKUYKLHYQKPL-NRFANRHFSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-(3-methylpentan-3-yloxy)-4-oxobutanoic acid
56777270
Solid
250mg
C25H29NO6
fmoc-asp ompe-oh, l-aspartic acid,n-9h-fluoren-9-ylmethoxy carbonyl-, 4-1-ethyl-1-methylpropyl ester, n-9h-fluoren-9-ylmethoxycarbonyl-l-aspartic acid 4-1-methyl-1-ethylpropyl ester, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-3-methylpentan-3-yl oxy-4-oxobutanoic acid non-preferred name, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-3-methylpentan-3-yl oxy-4-oxobutanoic acid
CCC(C)(CC)OC(=O)CC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
439.508
439.51
98%
N-Fmoc-L-aspartic acid 4-methylpentyl ester
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C