Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-aspartic acid 4-cyclohexyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Fmoc-L-aspartic acid 4-cyclohexyl ester
CAS 130304-80-2
Formula Weight 437.49
MDL Number MFCD00135420
Synonym Fmoc-Asp(OcHex)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6663906

 
alfa aesar™
H6663906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6663914

 
alfa aesar™
H6663914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 437.49
CAS 130304-80-2
Assay 95%
Physical Form Solid
Quantity 5g
MDL Number MFCD00135420
Chemical Name or Material N-Fmoc-L-aspartic acid 4-cyclohexyl ester
Storage Note 1 -30° to -10°C
Synonym Fmoc-Asp(OcHex)-OH
Safety and Handling

-30° to -10°C