missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-L-aspartic acid 1-benzyl ester, 95%, Thermo Scientific Chemicals
$86.60 - $310.25
Chemical Identifiers
CAS | 86060-83-5 |
---|---|
Molecular Formula | C26H23NO6 |
Molecular Weight (g/mol) | 445.471 |
MDL Number | MFCD00198201 |
InChI Key | CBZSVHFNEMONDZ-QHCPKHFHSA-N |
Synonym | fmoc-asp-obzl, a-benzyl ester, s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy-4-oxobutanoic acid, fmoc-l-aspartic acid-1-benzyl ester, fmoc-l-aspartic acid-1-benzyl ester, 3s-4-benzyloxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-l-asp-obzl, fmoc-aspartic acid-obzl, fmoc-aspartic acid-obzl, pubchem14955, fmoc-l-aspartic |
PubChem CID | 11224591 |
IUPAC Name | (3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-phenylmethoxybutanoic acid |
SMILES | C1=CC=C(C=C1)COC(=O)C(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6655803
|
Thermo Scientific Chemicals
H6655803 |
1 g |
Each for $86.60
|
|
|||||
AAH6655806
|
Thermo Scientific Chemicals
H6655806 |
5 g |
Each for $310.25
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
86060-83-5 | |
445.471 | |
CBZSVHFNEMONDZ-QHCPKHFHSA-N | |
11224591 | |
C1=CC=C(C=C1)COC(=O)C(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
C26H23NO6 | |
MFCD00198201 | |
fmoc-asp-obzl, a-benzyl ester, s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy-4-oxobutanoic acid, fmoc-l-aspartic acid-1-benzyl ester, fmoc-l-aspartic acid-1-benzyl ester, 3s-4-benzyloxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-l-asp-obzl, fmoc-aspartic acid-obzl, fmoc-aspartic acid-obzl, pubchem14955, fmoc-l-aspartic | |
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-phenylmethoxybutanoic acid |
Specifications
86060-83-5 | |
C26H23NO6 | |
4606552 | |
CBZSVHFNEMONDZ-QHCPKHFHSA-N | |
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-oxo-4-phenylmethoxybutanoic acid | |
11224591 | |
95% | |
Powder |
White | |
MFCD00198201 | |
fmoc-asp-obzl, a-benzyl ester, s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-benzyloxy-4-oxobutanoic acid, fmoc-l-aspartic acid-1-benzyl ester, fmoc-l-aspartic acid-1-benzyl ester, 3s-4-benzyloxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-l-asp-obzl, fmoc-aspartic acid-obzl, fmoc-aspartic acid-obzl, pubchem14955, fmoc-l-aspartic | |
C1=CC=C(C=C1)COC(=O)C(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
445.471 | |
445.47 | |
1 g | |
N-Fmoc-L-aspartic acid 1-benzyl ester |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only