Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-alpha-phenylglycine, 98%

$33.91 - $33.91

Chemical Identifiers

Chemical Name or Material N-Fmoc-L-alpha-phenylglycine
CAS 102410-65-1
Formula Weight 373.4
MDL Number MFCD00155632
Beilstein 5309005
Synonym Fmoc-Phg-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6301706

  SDS
alfa aesar™
H6301706
5g Each for $33.91

AAH6301714

  SDS
alfa aesar™
H6301714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 373.4
Synonym Fmoc-Phg-OH
MDL Number MFCD00155632
Chemical Name or Material N-Fmoc-L-alpha-phenylglycine
Quantity 5g
Beilstein 5309005
Storage Note 1 2° to 8°C
Assay 98%
CAS 102410-65-1
Physical Form Solid
Safety and Handling

2° to 8°C