missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-alpha-phenylglycine, 98%

Manufacturer:  Alfa Aesar™ H6301714

 View more versions of this product

Catalog No. AAH6301714



Specifications

Specifications

N-Fmoc-L-alpha-phenylglycine
C23H19NO4
5309005
PCJHOCNJLMFYCV-NRFANRHFSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2-phenylacetic acid
7269367
Solid
25g
102410-65-1
MFCD00155632
fmoc-phg-oh, fmoc-l-phenylglycine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-phenylacetic acid, n-fmoc-l-2-phenylglycine, fmoc-s-phenylglycine, n-9h-fluoren-9-ylmethoxy carbonyl-l-2-phenylglycine, n-alpha-9-fluorenylmethyloxycarbonyl-l-phenylglycine, s-9h-fluoren-9-ylmethoxycarbonylamino-phenyl-acetic acid, s-9h-fluoren-9-ylmethoxy carbonyl amino phenyl acetic acid, 2s-9h-fluoren-9-yl-methoxy carbonyl amino phenyl acetic acid
C1=CC=C(C=C1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
373.408
373.4
98%

Chemical Identifiers

102410-65-1
373.408
PCJHOCNJLMFYCV-NRFANRHFSA-N
7269367
C1=CC=C(C=C1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C23H19NO4
MFCD00155632
fmoc-phg-oh, fmoc-l-phenylglycine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-phenylacetic acid, n-fmoc-l-2-phenylglycine, fmoc-s-phenylglycine, n-9h-fluoren-9-ylmethoxy carbonyl-l-2-phenylglycine, n-alpha-9-fluorenylmethyloxycarbonyl-l-phenylglycine, s-9h-fluoren-9-ylmethoxycarbonylamino-phenyl-acetic acid, s-9h-fluoren-9-ylmethoxy carbonyl amino phenyl acetic acid, 2s-9h-fluoren-9-yl-methoxy carbonyl amino phenyl acetic acid
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-2-phenylacetic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents