Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-alaninol, 95%

Chemical Identifiers

Chemical Name or Material N-Fmoc-L-alaninol
CAS 161529-13-1
Formula Weight 297.35
MDL Number MFCD00235913
Synonym Fmoc-Ala-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6643303

 
alfa aesar™
H6643303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6643306

 
alfa aesar™
H6643306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 297.35
Chemical Name or Material N-Fmoc-L-alaninol
CAS 161529-13-1
Physical Form Solid
Assay 95%
Synonym Fmoc-Ala-ol
MDL Number MFCD00235913
Storage Note 1 -30° to -10°C
Quantity 1g
Safety and Handling

-30° to -10°C