Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-L-alanine N-succinimidyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Fmoc-L-alanine N-succinimidyl ester
CAS 73724-40-0
Formula Weight 408.41
MDL Number MFCD00065613
Beilstein 5462746
Synonym Fmoc-Ala-OSu; N-Fmoc-L-alanine N-hydroxysuccinimide ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6615803

 
alfa aesar™
H6615803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6615806

 
alfa aesar™
H6615806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6615814

 
alfa aesar™
H6615814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 408.41
Chemical Name or Material N-Fmoc-L-alanine N-succinimidyl ester
Physical Form Powder
Assay 95%
MDL Number MFCD00065613
Beilstein 5462746
Synonym Fmoc-Ala-OSu; N-Fmoc-L-alanine N-hydroxysuccinimide ester
Color Off-white to white
CAS 73724-40-0
Quantity 1g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C