missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-L-alanine monohydrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2110714
Description
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications
N-Fmoc-L-alanine monohydrate | |
∼130°C | |
MFCD00037139 | |
2225975 | |
Insoluble in water. | |
−19° (c=1 in DMF) | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid;hydrate | |
18528118 | |
98% |
207291-76-7 | |
C18H19NO5 | |
25 g | |
n-fmoc-l-alanine hydrate, n-fmoc-l-alanine monohydrate, n-9h-fluoren-9-ylmethoxy carbonyl-l-alanine hydrate, n-fmoc-l-alanine, 2s-2-fluoren-9-ylmethoxy carbonylamino propanoic acid, hydrate, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid hydrate, fmoc-ala-oh h2o, fmoc-ala-oh.h2o, fmoc-l-alanine hydrate, n-9-fluorenylmethoxycarbonyl-l-alanine monohydrate | |
GAPWKFLOMOFHGO-UHFFFAOYNA-N | |
O.CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
329.35 | |
329.36 (311.34 Anhydrous) |
Chemical Identifiers
207291-76-7 | |
329.35 | |
GAPWKFLOMOFHGO-UHFFFAOYNA-N | |
18528118 | |
O.CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
C18H19NO5 | |
MFCD00037139 | |
n-fmoc-l-alanine hydrate, n-fmoc-l-alanine monohydrate, n-9h-fluoren-9-ylmethoxy carbonyl-l-alanine hydrate, n-fmoc-l-alanine, 2s-2-fluoren-9-ylmethoxy carbonylamino propanoic acid, hydrate, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid hydrate, fmoc-ala-oh h2o, fmoc-ala-oh.h2o, fmoc-l-alanine hydrate, n-9-fluorenylmethoxycarbonyl-l-alanine monohydrate | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid;hydrate |
Safety and Handling
EINECSNumber : 252-660-6
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only