Please login to your online account to display your discounted pricing

N-FMOC-L-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-FMOC-L-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
CAS 136030-33-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C25 H21 N O4
Formula Weight 399.45
MDL Number MFCD00144368
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC301922500

  SDS
thermo fisher scientific
301922500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 399.45
Physical Form Granular Crystalline Powder
Quantity 250mg
Color Beige
CAS Max % 100.0
Assay Percent Range 99%
CAS Min % 98.5
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 136030-33-6
MDL Number MFCD00144368
Molecular Formula C25 H21 N O4
Chemical Name or Material N-FMOC-L-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid