missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-glycyl-L-valine, 95%

$86.04 - $1,138.08

Chemical Identifiers

CAS 86895-14-9
Molecular Formula C22H24N2O5
Molecular Weight (g/mol) 396.443
MDL Number MFCD00190884
InChI Key ORVHFHUXDLSLMX-FQEVSTJZSA-N
Synonym fmoc-gly-val-oh, l-valine, n-n-9h-fluoren-9-ylmethoxy carbonyl glycyl-9ci, fmoc-gly-val-oh, 2s-2-2-9h-fluoren-9-ylmethoxy carbonyl amino acetamido-3-methylbutanoic acid
PubChem CID 7019073
IUPAC Name (2S)-2-[[2-(9H-fluoren-9-ylmethoxycarbonylamino)acetyl]amino]-3-methylbutanoic acid
SMILES CC(C)C(C(=O)O)NC(=O)CNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH65664MD View Documents Alfa Aesar™
H65664MD
250mg Each for $86.04
 
AAH6566403 View Documents Alfa Aesar™
H6566403
1g Each for $272.96
 
AAH6566406 View Documents Alfa Aesar™
H6566406
5g Each for $1,138.08
 
Specifications

Chemical Identifiers

86895-14-9
396.443
ORVHFHUXDLSLMX-FQEVSTJZSA-N
7019073
CC(C)C(C(=O)O)NC(=O)CNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C22H24N2O5
MFCD00190884
fmoc-gly-val-oh, l-valine, n-n-9h-fluoren-9-ylmethoxy carbonyl glycyl-9ci, fmoc-gly-val-oh, 2s-2-2-9h-fluoren-9-ylmethoxy carbonyl amino acetamido-3-methylbutanoic acid
(2S)-2-[[2-(9H-fluoren-9-ylmethoxycarbonylamino)acetyl]amino]-3-methylbutanoic acid

Specifications

86895-14-9
MFCD00190884
fmoc-gly-val-oh, l-valine, n-n-9h-fluoren-9-ylmethoxy carbonyl glycyl-9ci, fmoc-gly-val-oh, 2s-2-2-9h-fluoren-9-ylmethoxy carbonyl amino acetamido-3-methylbutanoic acid
CC(C)C(C(=O)O)NC(=O)CNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
396.443
396.44
95%
N-Fmoc-glycyl-L-valine
C22H24N2O5
6829030
ORVHFHUXDLSLMX-FQEVSTJZSA-N
(2S)-2-[[2-(9H-fluoren-9-ylmethoxycarbonylamino)acetyl]amino]-3-methylbutanoic acid
7019073
Solid
250mg