missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-tyrosine, 97%

Chemical Identifiers

CAS 112883-29-1
Formula Weight 403.43
MDL Number MFCD00171384
Synonym Fmoc-D-Tyr-OH
Chemical Name or Material N-Fmoc-D-tyrosine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6674003 SDS Alfa Aesar™
H6674003
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6674006 SDS Alfa Aesar™
H6674006
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH66740MD SDS Alfa Aesar™
H66740MD
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

112883-29-1
MFCD00171384
N-Fmoc-D-tyrosine
403.43
Fmoc-D-Tyr-OH

Specifications

112883-29-1
403.43
Fmoc-D-Tyr-OH
175° to 180°C
N-Fmoc-D-tyrosine
Solid
MFCD00171384
White
1g
97%

Safety and Handling

storageNote1 : -30° to -10°C