missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-D-tyrosine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6674006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
175°C to 180°C | |
112883-29-1 | |
C24H21NO5 | |
fmoc-d-tyrosine, fmoc-d-tyr-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-hydroxyphenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-hydroxyphenyl propanoic acid, n-fmoc-d-tyr oh, fmoc-d-phe 4-oh-oh, n-9h-fluorene-9-ylmethoxycarbonyl-d-tyrosine, n-alpha-9-fluorenylmethoxycarbonyl-d-tyrosine, r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-4-hydroxy-phenyl-propionic acid | |
OC(=O)[C@@H](CC1=CC=C(O)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2 | |
403.43 | |
403.43 | |
5 g |
N-Fmoc-D-tyrosine | |
White | |
MFCD00171384 | |
SWZCTMTWRHEBIN-ANBDAQEENA-N | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-hydroxyphenyl)propanoic acid | |
6957988 | |
97% | |
Solid |
Chemical Identifiers
112883-29-1 | |
403.43 | |
SWZCTMTWRHEBIN-ANBDAQEENA-N | |
6957988 | |
OC(=O)[C@@H](CC1=CC=C(O)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2 |
C24H21NO5 | |
MFCD00171384 | |
fmoc-d-tyrosine, fmoc-d-tyr-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-hydroxyphenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-hydroxyphenyl propanoic acid, n-fmoc-d-tyr oh, fmoc-d-phe 4-oh-oh, n-9h-fluorene-9-ylmethoxycarbonyl-d-tyrosine, n-alpha-9-fluorenylmethoxycarbonyl-d-tyrosine, r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-4-hydroxy-phenyl-propionic acid | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-hydroxyphenyl)propanoic acid |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only