missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-tryptophan pentafluorophenyl ester, 97%

$28.84 - $337.73

Chemical Identifiers

CAS 136554-94-4
Molecular Formula C32H21F5N2O4
Molecular Weight (g/mol) 592.522
MDL Number MFCD00155634
InChI Key KLPGTAYCHANVFY-XMMPIXPASA-N
Synonym fmoc-d-trp-opfp, d-tryptophan,n-9h-fluoren-9-ylmethoxy carbonyl-, 2,3,4,5,6-pentafluorophenyl ester, d-tryptophan,n-9h-fluoren-9-ylmethoxy carbonyl-,2,3,4,5,6-pentafluorophenyl ester, 2,3,4,5,6-pentafluorophenyl 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1h-indol-3-yl propanoate
PubChem CID 53229920
IUPAC Name (2,3,4,5,6-pentafluorophenyl) (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1H-indol-3-yl)propanoate
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CNC5=CC=CC=C54)C(=O)OC6=C(C(=C(C(=C6F)F)F)F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6625703 View Documents Alfa Aesar™
H6625703
1g Each for $85.14
 
AAH6625706 View Documents Alfa Aesar™
H6625706
5g Each for $337.73
 
AAH66257MD View Documents Alfa Aesar™
H66257MD
250mg Each for $28.84
 
Specifications

Chemical Identifiers

136554-94-4
592.522
KLPGTAYCHANVFY-XMMPIXPASA-N
53229920
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CNC5=CC=CC=C54)C(=O)OC6=C(C(=C(C(=C6F)F)F)F)F
C32H21F5N2O4
MFCD00155634
fmoc-d-trp-opfp, d-tryptophan,n-9h-fluoren-9-ylmethoxy carbonyl-, 2,3,4,5,6-pentafluorophenyl ester, d-tryptophan,n-9h-fluoren-9-ylmethoxy carbonyl-,2,3,4,5,6-pentafluorophenyl ester, 2,3,4,5,6-pentafluorophenyl 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1h-indol-3-yl propanoate
(2,3,4,5,6-pentafluorophenyl) (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1H-indol-3-yl)propanoate

Specifications

136554-94-4
MFCD00155634
KLPGTAYCHANVFY-XMMPIXPASA-N
(2,3,4,5,6-pentafluorophenyl) (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1H-indol-3-yl)propanoate
53229920
Solid
1g
C32H21F5N2O4
fmoc-d-trp-opfp, d-tryptophan,n-9h-fluoren-9-ylmethoxy carbonyl-, 2,3,4,5,6-pentafluorophenyl ester, d-tryptophan,n-9h-fluoren-9-ylmethoxy carbonyl-,2,3,4,5,6-pentafluorophenyl ester, 2,3,4,5,6-pentafluorophenyl 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1h-indol-3-yl propanoate
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CNC5=CC=CC=C54)C(=O)OC6=C(C(=C(C(=C6F)F)F)F)F
592.522
592.52
97%
N-Fmoc-D-tryptophan pentafluorophenyl ester
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C