Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-threoninol, 98%

Chemical Identifiers

Chemical Name or Material N-Fmoc-D-threoninol
CAS 252049-02-8
Formula Weight 327.38
MDL Number MFCD00270219
Synonym Fmoc-D-Thr-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6695603

 
alfa aesar™
H6695603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6695606

 
alfa aesar™
H6695606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 327.38
CAS 252049-02-8
Chemical Name or Material N-Fmoc-D-threoninol
MDL Number MFCD00270219
Physical Form Solid
Assay 98%
Storage Note 1 2° to 8°C
Synonym Fmoc-D-Thr-ol
Quantity 1g
Safety and Handling

2° to 8°C