missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-serine, 95%

Manufacturer:  Alfa Aesar™ H6249303

 View more versions of this product

Catalog No. AAH6249303



Specifications

Specifications

N-Fmoc-D-serine
108°C to 112°C
116861-26-8
MFCD00077068
fmoc-d-ser-oh, fmoc-d-serine, n-9h-fluoren-9-ylmethoxy carbonyl-d-serine, n-fmoc-d-serine, d-serine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-serine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxypropanoic acid, ambotzfaa1508, pubchem10038, n-fmoc-d-ala oh
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CO)C(=O)O
327.336
327.33
+12° (c=1 in DMF)
1g
C18H17NO5
4715790
JZTKZVJMSCONAK-MRXNPFEDSA-N
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-hydroxypropanoic acid
688637
95%

Chemical Identifiers

116861-26-8
327.336
JZTKZVJMSCONAK-MRXNPFEDSA-N
688637
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CO)C(=O)O
C18H17NO5
MFCD00077068
fmoc-d-ser-oh, fmoc-d-serine, n-9h-fluoren-9-ylmethoxy carbonyl-d-serine, n-fmoc-d-serine, d-serine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-serine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxypropanoic acid, ambotzfaa1508, pubchem10038, n-fmoc-d-ala oh
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-hydroxypropanoic acid
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C

Documents