Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-proline, 98+%, may cont. up to ca 5% water

$51.38 - $577.09

Chemical Identifiers

Chemical Name or Material N-Fmoc-D-proline
CAS 101555-62-8
Molecular Formula C20H19NO4
Formula Weight 337.38
MDL Number MFCD00077067
Synonym N-(9-Fluorenylmethoxy)carbonyl-D-proline; Fmoc-D-Pro-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2850003

  SDS
alfa aesar™
H2850003
1g Each for $51.38

AAH2850006

  SDS
alfa aesar™
H2850006
5g Each for $150.68

AAH2850014

  SDS
alfa aesar™
H2850014
25g Each for $577.09
Description & Specifications

Specifications

Formula Weight 337.38
CAS 101555-62-8
Quantity 1g
Synonym N-(9-Fluorenylmethoxy)carbonyl-D-proline; Fmoc-D-Pro-OH
Recommended Storage 2° to 8°C
Chemical Name or Material N-Fmoc-D-proline
Melting Point 110° to 116°C
Molecular Formula C20H19NO4
Assay Percent Notes may contain up to ≈5% water
MDL Number MFCD00077067
Assay ≥98%

Applications
In organic synthesis, N-Fmoc-D-proline are often employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis, medicinal chemistry, and polymer chemistry.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

2° to 8°C