missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-pipecolic acid, ≥96%

Manufacturer:  Alfa Aesar™ H5745703

 View more versions of this product

Catalog No. AAH5745703



Specifications

Specifications

N-Fmoc-D-pipecolic acid
154°C to 159°C
101555-63-9
MFCD00235899
CKLAZLINARHOTG-LJQANCHMSA-N
(2R)-1-(9H-fluoren-9-ylmethoxycarbonyl)piperidine-2-carboxylic acid
6958378
Solid
Brown to White
1g
C21H21NO4
fmoc-d-pipecolic acid, fmoc-d-pip-oh, fmoc-d-hopro-oh, d-1-fmoc-pipecolinic acid, fmoc-d-piperidine-2-carboxylic acid, fmoc-d-homopro-oh, d-n-fmoc-pipecolic acid, r-n-fmoc-piperidine-2-carboxylic acid, r-1-9h-fluoren-9-yl methoxy carbonyl piperidine-2-carboxylic acid, fmoc-d-homoproline
C1CCN(C(C1)C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
351.402
351.4
≥96%

Chemical Identifiers

101555-63-9
351.402
CKLAZLINARHOTG-LJQANCHMSA-N
6958378
C1CCN(C(C1)C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C21H21NO4
MFCD00235899
fmoc-d-pipecolic acid, fmoc-d-pip-oh, fmoc-d-hopro-oh, d-1-fmoc-pipecolinic acid, fmoc-d-piperidine-2-carboxylic acid, fmoc-d-homopro-oh, d-n-fmoc-pipecolic acid, r-n-fmoc-piperidine-2-carboxylic acid, r-1-9h-fluoren-9-yl methoxy carbonyl piperidine-2-carboxylic acid, fmoc-d-homoproline
(2R)-1-(9H-fluoren-9-ylmethoxycarbonyl)piperidine-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

SDS Documents