Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-pipecolic acid, ≥96%

$20.20 - $20.20

Chemical Identifiers

Chemical Name or Material N-Fmoc-D-pipecolic acid
CAS 101555-63-9
Formula Weight 351.4
MDL Number MFCD00235899
Synonym (R)-(+)-1-Fmoc-piperidine-2-carboxylic acid; Fmoc-D-Pip-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5745703

  SDS
alfa aesar™
H5745703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH57457MD

  SDS
alfa aesar™
H57457MD
250mg Each for $20.20
Description & Specifications

Specifications

Formula Weight 351.4
Melting Point 154° to 159°C
Chemical Name or Material N-Fmoc-D-pipecolic acid
Physical Form Solid
Assay ≥96%
MDL Number MFCD00235899
CAS 101555-63-9
Synonym (R)-(+)-1-Fmoc-piperidine-2-carboxylic acid; Fmoc-D-Pip-OH
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
Color White to pale brown
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C