Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Fmoc-D-phenylalanine, 98%

$39.62 - $420.39

Chemical Identifiers

Chemical Name or Material N-Fmoc-D-phenylalanine
CAS 86123-10-6
Molecular Formula C24H21NO4
Formula Weight 387.44
MDL Number MFCD00062955
Synonym N-(9-Fluorenylmethoxycarbonyl)-D-phenylalanine; Fmoc-D-Phe-OH
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAB2168903

  SDS
alfa aesar™
B2168903
Each for $39.62

AAB2168906

  SDS
alfa aesar™
B2168906
Each for $132.59

AAB2168914

  SDS
alfa aesar™
B2168914
Each for $420.39
Description & Specifications

Specifications

Formula Weight 387.44
Recommended Storage 2° to 8°C
Assay 98%
MDL Number MFCD00062955
Synonym N-(9-Fluorenylmethoxycarbonyl)-D-phenylalanine; Fmoc-D-Phe-OH
Quantity 1g
Optical Rotation +40° (c=1 in DMF)
CAS 86123-10-6
Melting Point 181° to 185°C
Molecular Formula C24H21NO4
Chemical Name or Material N-Fmoc-D-phenylalanine
Safety and Handling

2° to 8°C